missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAS1R2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA534254
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Percent identity with other species by BLAST analysis: Human (100%) Chimpanzee, Gorilla, Orangutan, Gibbon (95%) Baboon, Monkey (85%).
TAS1R2 Polyclonal antibody specifically detects TAS1R2 in Human samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
TAS1R2 | |
Polyclonal | |
Unconjugated | |
TAS1R2 | |
aldehyde dehydrogenase 4 family, member A1; Aldh4a1; candidate taste receptor T1R2; G protein-coupled receptor 71; GPCRB4; GPR71; G-protein coupled receptor 71; Sweet taste receptor T1R2; T1R2; Tas1r2; taste 1 receptor member 2; taste receptor TR2; taste receptor type 1 member 2; taste receptor, type 1, member 2; Tr2; TS1R2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
80834 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q8TE23 | |
TAS1R2 | |
Synthetic 20 amino acid peptide from N-terminal extracellular domain of human TAS1R2. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |