missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAS2R40 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen OSR00164W
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Reconstitute with 100 μL of distilled water. Centrifuge to remove any insoluble material.
Gustducin-coupled receptor implicated in the perception of bitter compounds in the oral cavity and the gastrointestinal tract. Signals through PLCB2 and the calcium-regulated cation channel TRPM5. Expressed in subsets of taste receptor cells of the tongue and exclusively in gustducin-positive cells. Several bitter taste receptors are expressed in a single taste receptor cell.Spezifikation
TAS2R40 | |
Polyclonal | |
TAS2R40 | |
Taste receptor type 2 member 40, T2R40, T2R58, G-protein coupled receptor 60, MGC126844, MGC126846 | |
Rabbit | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
P59535 | |
TAS2R40 | |
A synthetic peptide from human taste receptor, type 2, member 40 | |
RUO | |
259286 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |