missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAS2R5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen OSR00154W
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Glycerol (1:1) may be added for additional stability. Reconstitute in 100 μL of sterile water.
This gene encodes a bitter taste receptor; bitter taste receptors are members of the G protein-coupled receptor superfamily and are specifically expressed by taste receptor cells of the tongue and palate epithelia. Each of these apparently intronless taste receptor genes encodes a 7-transmembrane receptor protein, functioning as a bitter taste receptor. This gene is clustered with another 3 candidate taste receptor genes on chromosome 7 and is genetically linked to loci that influence bitter perception. Receptor that may play a role in the perception of bitterness and is gustducin-linked. May play a role in sensing the chemical composition of the gastrointestinal content. The activity of this receptor may stimulate alpha gustducin, mediate PLC-beta-2 activation and lead to the gating of TRPM5. Expressed in subsets of taste receptor cells of the tongue and palate epithelium and exclusively in gustducin-positive cells. Most taste cells may be activated by a limited number of bitter compounds; individual taste cells can discriminate among bitter stimuli.Spezifikation
TAS2R5 | |
Polyclonal | |
TAS2R5 | |
Taste receptor type 2 member 5, T2R5, TAS2R5 | |
Rabbit | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Q9NYW4 | |
TAS2R5 | |
Synthetic peptide from human taste receptor, type 2, member 5. | |
RUO | |
54429 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |