missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAUT Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA537460
Weitere Details : Netto-Gewicht : 0.00910kg
Beschreibung
This antibody detects endogenous protein at a molecular weight of 70 kDa. Purity is >95% by SDS-PAGE.
TAUT Polyclonal antibody specifically detects TAUT in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
TAUT | |
Polyclonal | |
Unconjugated | |
SLC6A6 | |
AA589629; C80501; MGC10619; MGC131729; OTTHUMP00000208177; OTTHUMP00000208180; SC6A6; Slc6a6; Sodium- and chloride-dependent taurine transporter; Solute carrier 6 ,member 6 (taurine transporter); solute carrier family 6 (neurotransmitter transporter), member 6; solute carrier family 6 (neurotransmitter transporter, taurine), member 6; solute carrier family 6 member 6; solute carrier family 6, member 6; Taurine Transporter; taurine/beta-alanine transporter; TAUT | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
21366, 29464, 6533 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
O35316, P31641, P31643 | |
SLC6A6 | |
Synthetic peptide corresponding to amino acids 570-620 of human TAUT | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |