Learn More
TBC1D15 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA531804
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: PC-3. Predicted reactivity: Mouse (88%), Chimpanzee (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TBC1D15 Polyclonal antibody specifically detects TBC1D15 in Human samples. It is validated for Western Blot.Spezifikation
TBC1D15 | |
Polyclonal | |
Unconjugated | |
TBC1D15 | |
4432405K22Rik; AU022358; AU023730; AW557774; GAP for RAB7; GTPase-activating protein RAB7; Ly6dl; lymphocyte antigen 6, locus D ligand; Rab7-GAP; TBC1 domain family member 15; TBC1 domain family, member 15; Tbc1d15; Tre2/Bub2/Cdc16 domain family member 15; Tre-2-budding uninhibited by benzimidazole-cell division cycle 16 domain, domain family member 15 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64786 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8TC07 | |
TBC1D15 | |
Recombinant fragment corresponding to a region within amino acids 1 and 190 of Human TBC1D15 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |