missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBCCD1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA524074
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
TBCCD1 Polyclonal antibody specifically detects TBCCD1 in Human, Mouse samples. It is validated for Western Blot.Spezifikation
TBCCD1 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q640P7, Q9NVR7 | |
TBCCD1 | |
KLH conjugated synthetic peptide between 491-520 amino acids from the C-terminal region of human TBCCD1 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
TBCCD1 | |
5730478M09Rik; AU015083; BB165076; TBCC domain containing 1; TBCC domain-containing protein 1; Tbccd1 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
55171, 70573 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |