missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBK1 Monoclonal Antibody (108A429), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA516159
Weitere Details : Netto-Gewicht : 0.10000kg
Beschreibung
Suggested positive control: human testis protein, NIH-3T3 or RAW.
TBK1 Monoclonal antibody specifically detects TBK1 in Bovine, Canine, Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Spezifikation
TBK1 | |
Monoclonal | |
1.0 mg/mL | |
PBS with 0.05% sodium azide | |
Q9UHD2, Q9WUN2 | |
TBK1 | |
Synthetic peptide (YYQFKKDKAERRLAYC ) of TBK1/NAK protein. | |
100 μg | |
Primary | |
Bovine, Canine, Human, Mouse, Rat | |
Antibody | |
IgG1 κ |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
108A429 | |
Unconjugated | |
TBK1 | |
1200008B05Rik; AI462036; AW048562; FLJ11330; FTDALS4; NAK; NF-kappa-B-activating kinase; NF-kB-activating kinase; serine/threonine protein kinase TBK1; serine/threonine-protein kinase TBK1; T2K; TANK binding kinase 1; TANK-binding kinase 1; tbk; TBK1 | |
Mouse | |
Protein G | |
RUO | |
29110, 299827, 481145, 533141, 56480 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |