Learn More
TBX20 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA540669
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: MEFTASPKPQ LSSRANAFSI AALMSSGGSK EKEATENTIK PLEQFVEKSS Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%
TBX20 Polyclonal antibody specifically detects TBX20 in Human samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
TBX20 | |
Polyclonal | |
Unconjugated | |
TBX20 | |
9430010M06Rik; AL022859; ASD4; cb826; H15-related T-box transcription factor hrT; hrt; T-box 20; T-box protein 20; T-box transcription factor; T-box transcription factor 20; T-box transcription factor TBX20; T-box transcription factor TBX20-like; Tbx12; TBX20; whz; ZF-TBX20 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
57057 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9UMR3 | |
TBX20 | |
Synthetic peptide directed towards the N-terminal of human TBX20 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |