missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBX3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen 424800
Weitere Details : Netto-Gewicht : 0.50000kg
Beschreibung
42-4800 has been successfully used in immunohistochemistry analysis of Tbx3 in mouse E16.5 frozen lung sections.
TBX3 Polyclonal antibody specifically detects TBX3 in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot.Spezifikation
TBX3 | |
Polyclonal | |
Unconjugated | |
Tbx3 | |
bladder cancer related protein XHL; D5Ertd189e; T-box 3; T-box 3 (ulnar mammary syndrome); T-box protein 3; T-box transcription factor TBX3; TBX3; TBX3-ISO; UMS; XHL | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
21386, 353305, 6926 | |
-20°C | |
Liquid |
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
0.25 mg/mL | |
PBS with 0.1% sodium azide; pH 7.4 | |
O15119, P70324, Q7TST9 | |
Tbx3 | |
Synthetic peptide derived from an internal region of human T-box 3, which differs from mouse and rat by one conservative and one non-conservative amino acid | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |