missing translation for 'onlineSavingsMsg'
Learn More

TCCR Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA519985

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 12898192

  • 514.00 € / 100 Mikrogramm
Verfügbar ab: 19-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

In Western blot applications, this antibody detects a band at ∽70kDa. A suggested positive control is human spleen tissue lysate. PA5-19985 can be used with blocking peptide PEP-0109.

TCCR Polyclonal antibody specifically detects TCCR in Human samples. It is validated for Immunohistochemistry, Western Blot.
Spezifikation

Spezifikation

TCCR
Polyclonal
Unconjugated
Il27ra
class I cytokine receptor; CRL1; CRL1IL-27R; cytokine receptor family, class 1 (WSXWS), member 1; cytokine receptor WSX-1; Cytokine receptor-like 1; il-27 receptor; IL-27 receptor subunit alpha; IL27R; IL-27R; IL-27R subunit alpha; IL27RA; IL-27RA; IL-27R-alpha; interleukin 27 receptor subunit alpha; interleukin 27 receptor, alpha; interleukin-27 receptor subunit alpha; T cell cytokine receptor; T cell cytokine receptor; cytokine receptor family, class 1 (WSXWS), member 1; TCCR; T-cell cytokine receptor type 1; Type I T-cell cytokine receptor; UNQ296/PRO336; WSX1; WSX-1; ZcytoR1
Rabbit
Antigen affinity chromatography
RUO
9466
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Liquid
Immunohistochemistry, Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q6UWB1
Il27ra
A synthetic peptide corresponding to amino acids near the carboxy terminus of human TCCR precursor
100 μg
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote