missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCEAL1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA536612
Weitere Details : Netto-Gewicht : 0.00910kg
Beschreibung
This antibody detects endogenous protein at a molecular weight of 18 kDa. Purity is >95% by SDS-PAGE.
TCEAL1 Polyclonal antibody specifically detects TCEAL1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
TCEAL1 | |
Polyclonal | |
Unconjugated | |
TCEAL1 | |
0610011M09Rik; BC011290; Nuclear phosphoprotein p21/SIIR; P21; pp21; SIIR; TCEAL1; TCEA-like protein 1; transcription elongation factor A (SII)-like 1; transcription elongation factor A like 1; transcription elongation factor A protein-like 1; Transcription elongation factor S-II protein-like 1; WEX9 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
237052, 302593, 9338 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q15170, Q5PPP3, Q921P9 | |
TCEAL1 | |
Synthetic peptide corresponding to amino acids 90-143 of human TCEAL1 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |