missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCF12 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA521039
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
A suggested positive control is Hela cell lysate. PA5-21039 can be used with blocking peptide PEP-1153.
TCF12 Polyclonal antibody specifically detects TCF12 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
TCF12 | |
Polyclonal | |
Unconjugated | |
TCF12 | |
A130037E08Rik; ALF1; bHLHb20; Class A helix-loop-helix transcription factor ME1; class B basic helix-loop-helix protein 20; CRS3; DNA-binding protein HTF4; E-box-binding protein; HEB; HEBAlt; helix-loop-helix transcription factor 4; HsT17266; HTF4; HTF-4; ME1; Meb; REB; Salivary-specific cAMP response element-binding protein alpha; SCBP alpha; SCBPA; SCBP-alpha; TCF12; TCF-12; Transcription factor 12; transcription factor HTF-4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
21406, 25720, 6938 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
P51514, Q61286, Q99081 | |
TCF12 | |
A 19 amino acid peptide near the amino terminus of human TCF12. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |