missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCF19 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA521744
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: 293T. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TCF19 Polyclonal antibody specifically detects TCF19 in Human samples. It is validated for Western Blot.Spezifikation
TCF19 | |
Polyclonal | |
Unconjugated | |
TCF19 | |
5730403J10Rik; AW495861; DAMA-213L4.5; SC 1; SC1; SC1 protein homolog; SC1(TCF19)-6; SC1(TCF19)-7; SC1-1; SCI(TCF19)-4; TCF 19; TCF19; TCF-19; Transcription factor 19; transcription factor 19-like protein; Transcription factor SC1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6941 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.96 mg/mL | |
PBS with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q9Y242 | |
TCF19 | |
Synthetic peptide contain a sequence corresponding to a region within amino acids 296 and 309 (Q9Y242) of TCF19 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |