missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCF7L1 Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Marke: Invitrogen PA547810
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Reconstitute at 0.2 mg/mL in sterile PBS.
TCF7L1 Polyclonal antibody specifically detects TCF7L1 in Human samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
TCF7L1 | |
Polyclonal | |
Unconjugated | |
TCF7L1 | |
bHLHb21; HMG box transcription factor 3; T-cell factor 3; Tcf3; TCF-3; TCF7L1; transcription factor 3; transcription factor 7 like 1; transcription factor 7 like 1 (T cell specific, HMG box); transcription factor 7-like 1; transcription factor 7-like 1 (T-cell specific, HMG box); transcription factor 7-like 1 (T-cell specific, HMG-box); transcription factor 7-like 2 (T-cell specific, HMG box) | |
Sheep | |
Antigen affinity chromatography | |
RUO | |
83439 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q9HCS4 | |
TCF7L1 | |
E. coli-derived recombinant human TCF7L1 Lys429-Ser581 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |