Learn More
TCP-1 theta Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA530403
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: 293T, A431, Jurkat, Raji. Predicted reactivity: Mouse (97%), Rat (98%), Zebrafish (84%), Xenopus laevis (87%), Chicken (95%), Rhesus Monkey (99%), Chimpanzee (98%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TCP-1 theta Polyclonal antibody specifically detects TCP-1 theta in Human samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
TCP-1 theta | |
Polyclonal | |
Unconjugated | |
CCT8 | |
AI132397; bdav; bette davis; C21orf112; Cct8; Cctq; CCT-theta; Chaperonin containing T-complex polypeptide 1 subunit 8; chaperonin containing TCP1 subunit 8; chaperonin containing TCP1, subunit 8 (theta); chaperonin subunit 8 (theta); D21S246; fa22h09; KIAA0002; PRED71; renal carcinoma antigen NY-REN-15; T-complex protein 1 subunit theta; TCP-1-theta; Tcpq; wu:fa22h09; zgc:56059 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10694 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.99 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
P50990 | |
CCT8 | |
Recombinant fragment corresponding to a region within amino acids 284 and 527 of Human TCP1 theta | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |