missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCR alpha/beta Monoclonal Antibody (IP26), APC, Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA110452
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody recognizes a monomorphic determinant of TCR alpha/beta.
TCR alpha/beta Monoclonal antibody specifically detects TCR alpha/beta in Human samples. It is validated for Flow Cytometry.Spezifikation
TCR alpha/beta | |
Monoclonal | |
APC | |
TRAC | |
FLJ22602; IMD7; LOC290071; MGC117436; MGC22624; MGC23964; MGC71411; RATTCB; RATTCBC1; RGD1359684; similar to RIKEN cDNA A430107P09 gene; T cell receptor beta locus; T3/TCR complex; TCB; TCBC1; t-cell antigen receptor; T-cell receptor alpha constant; T-cell receptor beta chain; T-cell receptor V alpha; tcr alpha; TCR alpha/ beta; TCR beta; TCRA; Tcrb; TRA; Tra29; TRAC; TRB; TRB@; TRCA | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
28589, 28755, 6955, 6957 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
IP26 | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
P01848, P04435 | |
TRA, TRAC, TRB, TRBV7-9 | |
not available | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |