Learn More
TCR V alpha 2 Monoclonal Antibody (F1), FITC, Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen TCR2663
Weitere Details : Netto-Gewicht : 0.00400kg Transport : UN number : 3261 Chem class : 8 Pack group : III
Beschreibung
The ability of T cell receptors (TCR) to discriminate foreign from self-peptides presented by major histocompatibility complex (MHC) class II molecules is essential for an effective adaptive immune response. TCR recognition of self-peptides has been linked to autoimmune disease. Mutant self-peptides have been associated with tumors. Engagement of TCRs by a family of bacterial toxins know as superantigens has been responsible for toxic shock syndrome. Autoantibodies to V beta segments of T cell receptors have been isolated from patients with rheumatoid arthritis (RA) and systemic lupus erythematosus (SLE). The autoantibodies block TH1-mediated inflammatory autodestructive reactions and are believed to be a method by which the immune system compensates for disease. Most human T cells express the TCR alpha-beta and either CD4 or CD8 molecule (single positive, SP). A small number of T cells lack both CD4 and CD8 (double negative, DN). Increased percentages of alpha-beta DN T cells have been identified in some autoimmune and immunodeficiency disorders. Gamma-delta T cells are primarily found within the epithelium. They show less TCR diversity and recognize antigens differently than alpha-beta T cells. Subsets of gamma-delta T cells have shown antitumor and immunoregulatory activity.Spezifikation
TCR V alpha 2 | |
Monoclonal | |
0.15 mg/mL | |
PBS with 0.5% BSA and 0.1% sodium azide | |
Mouse | |
Protein A | |
RUO | |
Human, Non-human Primate | |
Antibody | |
IgG2a |
Flow Cytometry | |
F1 | |
FITC | |
TCRVA; TCRVA2; V alpha 2; V alpha-2 TCR; Va 2; Va2; Valpha 2; Valpha2 | |
Human TCR V alpha-2 | |
200 Tests | |
Primary | |
4° C, store in dark | |
Liquid |