missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRPM1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen OSR00048W
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Reconstitute with 100 μL of distilled water. Centrifuge to remove any insoluble material. OSR00048W can be used with blocking peptide GST00008P.
The protein encoded by this gene is similar to the transient receptor potential (TRP) calcium channel family members. The expression of this protein is inversely correlated with melanoma aggressiveness, suggesting that it suppresses melanoma metastasis.Spezifikation
TRPM1 | |
Polyclonal | |
Trpm1 | |
Transient receptor potential cation channel,subfamily M, member 1 variant delta1066-1071, Trpm1, LTRPC1, MLSN1, melastatin | |
Rabbit | |
100 μL | |
Primary | |
Rat | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Q2WEA5 | |
TRPM1 | |
A synthetic peptide from rat TRPM1 conjugated to an immunogenic carrier protein | |
RUO | |
361586 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |