missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSC22D2 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA535176
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
TSC22D2 belongs to the TSC-22/Dip/Bun family and the function remains unknown.Spezifikation
TSC22D2 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
O75157 | |
TSC22D2 | |
KLH conjugated synthetic peptide between 34-62 amino acids from the N-terminal region of human TSC22D2 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
TSC22D2 | |
TSC22D2; KIAA0669; TILZ4; TSC22 domain family protein 2; TSC22-related-inducible leucine zipper protein 4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9819 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |