missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GILZ Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA534506
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
A suggested positive control is human small intestine tissue lysate. PA5-34506 can be used with blocking peptide PEP-1549.
GILZ Polyclonal antibody specifically detects GILZ in Human, Mouse samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
GILZ | |
Polyclonal | |
Unconjugated | |
TSC22D3 | |
delta sleep inducing peptide, immunoreactor; delta sleep-inducing peptide immunoreactor; DIP; Dsip1; DSIPI; DSIP-immunoreactive leucine zipper protein; DSIP-immunoreactive peptide; GILZ; glucocorticoid-induced leucine zipper protein; long glucocorticoid-induced leucine zipper protein; Protein DIP; Tilz3; TSC22 domain family 3; TSC22 domain family member 3; TSC22 domain family protein 3; TSC22 domain family, member 3; TSC-22 related protein; TSC22D3; TSC-22-like protein; TSC-22R; TSC22-related inducible leucine zipper 3; TSC-22-related protein; TSC22-related-inducible leucine zipper 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
14605, 1831 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q99576, Q9Z2S7 | |
TSC22D3 | |
A 16 amino acid peptide near the amino terminus of human TSC22D3 | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |