Learn More
TSFM Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA527652
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: Jurkat, Raji, K562. Predicted reactivity: Mouse (89%), Rat (89%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TSFM Polyclonal antibody specifically detects TSFM in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
TSFM | |
Polyclonal | |
Unconjugated | |
TSFM | |
2310050B20Rik; 2A3-2; 9430024O13Rik; EFTS; EF-Ts; EF-Ts {ECO:0000255; EFTSMT; EF-TsMt; EF-TsMt {ECO:0000255; elongation factor Ts; elongation factor Ts, mitochondrial; elongation factor Ts, mitochondrial {ECO:0000255; HAMAP-Rule:MF_03135}; mitochondrial elongation factor Ts; translation elongation factor; Ts translation elongation factor, mitochondrial; TSFM; zgc:158429 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10102 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
P43897 | |
TSFM | |
Recombinant protein fragment corresponding to a region within amino acids 128 and 275 of Human TSFM | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |