missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSG101 Monoclonal Antibody (4A10), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA123296
Weitere Details : Netto-Gewicht : 1.24740kg
Beschreibung
Suggested positive controls for this product are MOLT4 and HeLa.
TSG101 Monoclonal antibody specifically detects TSG101 in Hamster, Human, Mouse, Non-human Primate, Rat samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunomicroscopy, Immunoprecipitation, Western Blot.Spezifikation
TSG101 | |
Monoclonal | |
1.8 mg/mL | |
PBS with no preservative | |
Q61187, Q6IRE4, Q99816 | |
Tsg101 | |
Residues 167-374 of TSG101, expressed in E. coli. | |
100 μL | |
Primary | |
Hamster, Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunomicroscopy, Immunoprecipitation, Western Blot | |
4A10 | |
Unconjugated | |
Tsg101 | |
AI255943; CC2; ESCRTI complex subunit TSG101; ESCRT-I complex subunit TSG101; Rw; TSG 101; TSG10; TSG101; Tumor supressor; tumor susceptibility 101; tumor susceptibility gene 10; tumor susceptibility gene 101; tumor susceptibility gene 101 protein; tumor susceptibility protein; VPS23 | |
Mouse | |
Affinity chromatography | |
RUO | |
100752541, 22088, 292925, 7251 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |