missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSGA2 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA518946
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with bovine, human and rat based on sequence homology.
TSGA2 Polyclonal antibody specifically detects TSGA2 in Mouse samples. It is validated for Western Blot.Spezifikation
TSGA2 | |
Polyclonal | |
Unconjugated | |
RSPH1 | |
ABCB2; cancer/testis antigen 79; CT79; h-meichroacidin; Male meiotic metaphase chromosome-associated acidic protein; MCA; Meichroacidin; MGC126568; MGC141927; PSF1; radial spoke head 1 homolog; radial spoke head 1 homolog (Chlamydomonas); RING4; RSP44; Rsph1; RSPH10A; testes specific gene A2 homolog; testis specific A2 homolog; testis specific gene A2; testis-specific gene A2 protein; TSA2; TSGA2; TSGA2 epididymal isoform | |
Goat | |
Antigen affinity chromatography | |
RUO | |
22092 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q8VIG3 | |
RSPH1 | |
Synthetic peptide sequence (HLNHRYQGKFLNKNP) corresponding to the internal amino acids of RSPH1 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |