Learn More
TSH Receptor Monoclonal Antibody (4C1), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA516519
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. A suggested positive control for immunohistochemical applications is thyroid gland. This product does not require protein digestion pre-treatment of paraffin embedded sections. This product does not require antigen retrieval using heat treatment prior to staining of paraffin embedded sections.
TSH Receptor Monoclonal antibody specifically detects TSH Receptor in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Spezifikation
TSH Receptor | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
P16473 | |
TSHR | |
External domain of human TSH receptor. | |
200 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
4C1 | |
Unconjugated | |
TSHR | |
AI481368; CHNG1; hTSHR-I; hypothroid; hyt; LGR3; MGC141920; pet; seven transmembrane helix receptor; thyroid stimulating hormone receptor; thyroid stimulating hormone receptor, isoform 2; thyroid-stimulating hormone receptor; thyroid-stimulating hormone receptor variant; thyrotropin receptor; thyrotropin receptor-I, hTSHR-I; TSHR; TSH-R; TSHR; seven transmembrane domain G protein-coupled receptor; TSHRA | |
Mouse | |
Protein A | |
RUO | |
7253 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |