missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thrombospondin 1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA129196
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Positive Control: HUVEC Cells
Thrombospondin 1 Polyclonal antibody specifically detects Thrombospondin 1 in Bovine, Human, Mouse samples. It is validated for Immunocytochemistry, Immunoprecipitation, Western Blot.Spezifikation
Thrombospondin 1 | |
Polyclonal | |
Unconjugated | |
THBS1 | |
glycoprotein G; tbsp1; THBS; Thbs1; Thbs-1; thrombospondin; thrombospondin 1; thrombospondin-1; thrombospondin-1p180; thrombospondin-p50; ThrombospondinTHBS1; TSP; TSP1; TSP-1 | |
Rabbit | |
Protein A | |
RUO | |
21825, 281530, 7057 | |
4° C, do not freeze | |
Liquid |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
P07996, P35441, Q28178 | |
THBS1 | |
Purified human platelet TSP | |
500 μL | |
Primary | |
Bovine, Human, Mouse | |
Antibody | |
IgG |