missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSPAN6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA533094
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Percent identity with other species by BLAST analysis: Human, Gorilla, Orangutan, Gibbon (100%) Dog, Horse, Pig (94%) Mouse, Bat, Elephant, Panda, Rabbit (88%).
TSPAN6 Polyclonal antibody specifically detects TSPAN6 in Human samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
TSPAN6 | |
Polyclonal | |
Unconjugated | |
TSPAN6 | |
6720473L21Rik; A15 homolog; AI316786; Putative NF-kappa-B-activating protein 321; T245; T245 protein; tetraspan TM4SF; tetraspanin 6; tetraspanin TM4-D; Tetraspanin-6; Tm4sf; TM4SF6; Transmembrane 4 superfamily member 6; TSPAN6; Tspan-6; UNQ767/PRO1560 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7105 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
O43657 | |
TSPAN6 | |
Synthetic 16 amino acid peptide from internal region of human TSPAN6. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |