missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSPY1L Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA534483
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
A suggested positive control is A-20 cell lysate. PA5-34483 can be used with blocking peptide PEP-1525.
TSPY1L Polyclonal antibody specifically detects TSPY1L in Human, Mouse samples. It is validated for Western Blot.Spezifikation
TSPY1L | |
Polyclonal | |
Unconjugated | |
TSPY1 | |
cancer/testis antigen 78; CT78; DYS14; pJA923; testis specific protein, Y-linked 1; testis-specific Y-encoded protein 1; TSPY; TSPY1; TSPY1L; TSPY1S | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7258 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q01534 | |
TSPY1 | |
A 20 amino acid peptide near the carboxy terminus of human TSPY1L | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |