Learn More
TTC1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA521373
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: 293T. Predicted reactivity: Rhesus Monkey (98%), Bovine (81%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TTC1 Polyclonal antibody specifically detects TTC1 in Human samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
TTC1 | |
Polyclonal | |
Unconjugated | |
Ttc1 | |
4833412C19Rik; C79328; tetratricopeptide repeat domain 1; tetratricopeptide repeat protein 1; TPR repeat protein 1; TPR1; Ttc1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7265 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q99614 | |
Ttc1 | |
Recombinant fragment corresponding to a region within amino acids 1 and 210 of TTC1 (Uniprot ID#Q99614) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |