missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTC31 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA532080
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: THP-1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TTC31 Polyclonal antibody specifically detects TTC31 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
TTC31 | |
Polyclonal | |
Unconjugated | |
TTC31 | |
tetratricopeptide repeat domain 31; tetratricopeptide repeat protein 31; TPR repeat protein 31; TTC31 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64427 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q49AM3 | |
TTC31 | |
Recombinant protein fragment corresponding to a region within amino acids 1 and 519 of Human TTC31 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |