missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTC8 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA519300
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with bovine, canine, mouse and porcine based on sequence homology.
TTC8 Polyclonal antibody specifically detects TTC8 in Human samples. It is validated for Western Blot.Spezifikation
TTC8 | |
Polyclonal | |
Unconjugated | |
TTC8 | |
0610012F22Rik; AV001447; Bardet-Biedl syndrome 8 protein; Bardet-Biedl syndrome 8 protein homolog; Bardet-Biedl syndrome type 8; BBS8; RP51; tetratricopeptide repeat domain 8; tetratricopeptide repeat protein 8; TPR repeat protein 8; TTC8 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
123016 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q8TAM2 | |
TTC8 | |
Synthetic peptide sequence (KEVLKQDNTHVE) corresponding to the internal amino acids of TTC8 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |