missing translation for 'onlineSavingsMsg'
Learn More

TTF2 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA529065

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 13443187

  • 473.00 € / 100 Mikroliter
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Recommended positive controls: H1299. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

TTF2 Polyclonal antibody specifically detects TTF2 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.
Spezifikation

Spezifikation

TTF2
Polyclonal
Unconjugated
TTF2
4632434F22Rik; 8030447N19; AV218430; F2; HuF2; human factor 2; lodestar homolog; lodestar protein; RNA polymerase II termination factor; Transcription release factor 2; transcription termination factor 2; transcription termination factor, RNA polymerase II; Ttf2; ZGRF6; zinc finger, GRF-type containing 6
Rabbit
Antigen affinity chromatography
RUO
8458
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.93 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q9UNY4
TTF2
Recombinant fragment corresponding to a region within amino acids 232 and 511 of Human TTF2
100 μL
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote