Learn More
TUB Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA531164
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: NT2D1, PC-3, U87-MG, SK-N-SH, Neuro2A, GL261. Predicted reactivity: Mouse (92%), Rat (93%), Bovine (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TUB Polyclonal antibody specifically detects TUB in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
TUB | |
Polyclonal | |
Unconjugated | |
TUB | |
CELE_F10B5.4; F10B5.4; Rd5; RDOB; retinal degeneration 5; TUB; TUB bipartite transcription factor; TUB-1; tubby bipartite transcription factor; tubby candidate gene; tubby homolog; tubby homologue; tubby protein; Tubby protein homolog; Tubby protein homolog 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22141, 25609, 7275 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
O88808, P50586, P50607 | |
TUB | |
Recombinant protein fragment corresponding to a region within amino acids 4 and 263 of Human Tub | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |