missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-3 Tubulin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA525655
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with bovine, chicken, non-human primate and rat based on sequence homology.
beta-3 Tubulin Polyclonal antibody specifically detects beta-3 Tubulin in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
beta-3 Tubulin | |
Polyclonal | |
Unconjugated | |
TUBB3 | |
3200002H15Rik; beta 3 tubulin; beta tubulin 3; beta-4; betaIII-tubulin; CDCBM; CDCBM1; CFEOM3; CFEOM3A; class III beta-tubulin; FEOM3; I79_000194; LOC100341581; M(beta)3; M(beta)6; mc1R; melanocortin receptor 1; Neuron specific beta III Tubulin; neuron-specific class III beta-tubulin; TUBB; Tubb3; TUBB4; tubulin beta; tubulin beta 3 class III; Tubulin beta-3; tubulin beta-3 chain; Tubulin beta-4; Tubulin beta-4 chain; tubulin beta-III; tubulin, beta 3 class III; tubulin, beta, 3; tubulin, beta, 4; Unknown (protein for MGC:134115) | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
10381, 22152, 246118 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 0.09% sodium azide | |
Q13509, Q4QRB4, Q9ERD7 | |
TUBB3 | |
KLH conjugated synthetic peptide between 36-63 amino acids from the N-terminal region of human TUBB3 | |
400 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |