missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSC2 Monoclonal Antibody (3G9D9), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen 370500
Weitere Details : Netto-Gewicht : 0.50000kg
Beschreibung
TSC2 Monoclonal antibody specifically detects TSC2 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Spezifikation
TSC2 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.1% sodium azide; pH 7.4 | |
P49815 | |
TSC2 | |
Recombinant human tuberin protein | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
3G9D9 | |
Unconjugated | |
TSC2 | |
LAM; Nafld; OTTHUMP00000198394; PPP1R160; protein phosphatase 1, regulatory subunit 160; Rc; renal carcinoma; Tcs2; Tsc2; TSC4; Tuberin; tuberous sclerosis 2; tuberous sclerosis 2 homolog protein; tuberous sclerosis 2 protein; Tuberous sclerosis 2 protein homolog | |
Mouse | |
Protein A | |
RUO | |
7249 | |
-20°C | |
Liquid |