missing translation for 'onlineSavingsMsg'
Learn More
Learn More
gamma Tubulin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA128042
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: chicken fibroblast, A431. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
gamma Tubulin Polyclonal antibody specifically detects gamma Tubulin in Chicken, Hamster, Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
gamma Tubulin | |
Polyclonal | |
Unconjugated | |
TUBG1 | |
1500010O08Rik; AI451582; AI503389; AI504772; CDCBM4; centrosome marker; gamma tubulin; gamma-1-tubulin; Gamma-2-tubulin; gamma-tubulin; gamma-tubulin complex component 1; GCP-1; similar to tubulin, gamma 1; Tubg; TUBG1; tubg1.L; TUBG2; TUBGCP1; Tubgl; tubulin gamma 1; tubulin gamma 1 L homeolog; tubulin gamma 2; Tubulin gamma-1 chain; Tubulin gamma-2 chain; tubulin, gamma 1; tubulin, gamma 2; tubulin, gamma polypeptide; Unknown (protein for MGC:133882); XELAEV_18002398mg; xgam; zgc:55894 | |
Rabbit | |
IgG fraction | |
RUO | |
100759432, 103733, 103768, 252921, 27175, 420025, 680991, 7283 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
10.5 mg/mL | |
0.01M PBS with 15mM sodium azide; pH 7.4 | |
P23258, P83887, P83888, Q8VCK3, Q9NRH3 | |
TUBG1, Tubg2 | |
synthetic peptide EEFATEGTDRKDVFFY-K conjugated to KLH, corresponding to amino acids 38-53 of Human gamma Tubulin. | |
100 μL | |
Primary | |
Chicken, Hamster, Human, Mouse, Rat | |
Antibody | |
IgG |