Learn More
Tug Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke:  Invitrogen PA517022
Weitere Details : Netto-Gewicht : 0.01000kg
Artikelnummer. 11551491
  • 422.00 € / 100 Mikroliter
Verfügbar ab: 22-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
It is not recommended to aliquot this antibody.
Tug Polyclonal antibody specifically detects Tug in Human, Mouse, Non-human Primate, Rat samples. It is validated for Immunoprecipitation, Western Blot.
Spezifikation
Spezifikation
Tug
Polyclonal
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5
Q8VBT9, Q9BZE9
ASPSCR1
Synthetic peptide corresponding to amino acids around residue Val542 of human Tug
100 μL
Primary
Human, Mouse, Non-human Primate, Rat
Antibody
IgG
Immunoprecipitation, Western Blot
Unconjugated
ASPSCR1
1190006K01Rik; alveolar soft part sarcoma chromosomal region candidate gene 1 protein; Alveolar soft part sarcoma chromosomal region candidate gene 1 protein homolog; alveolar soft part sarcoma chromosome region, candidate 1; alveolar soft part sarcoma chromosome region, candidate 1 (human); alveolar soft part sarcoma locus; ASPC; ASPCR1; ASPL; ASPS; Aspscr1; ASPSCR1, UBX domain containing tether for SLC2A4; RCC17; renal cell carcinoma gene on chromosome 17; renal cell carcinoma, papillary, 17; renal papillary cell carcinoma protein 17; Tether containing UBX domain for GLUT4; tether, containing a UBX domain, for GLUT4; TUG; UBX domain protein 9; UBX domain-containing protein 9; UBXD9; UBXN9
Rabbit
Antigen affinity chromatography
RUO
68938, 691026, 79058
-20°C