Learn More
CD266 Monoclonal Antibody (314636), Invitrogen™
Rat Monoclonal Antibody
Marke: Invitrogen MA524037
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse (rm) EDA, rmNeogenin, recombinant human TWEAK R, rmOPG, or rmTNF R1 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
CD266 Monoclonal antibody specifically detects CD266 in Mouse samples. It is validated for Flow Cytometry, Western Blot.Spezifikation
CD266 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q9CR75 | |
TNFRSF12A | |
E. coli-derived recombinant mouse TWEAK R Glu28-Trp79 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a |
Flow Cytometry, Western Blot | |
314636 | |
Unconjugated | |
TNFRSF12A | |
AI255180; C87282; CD266; FGF-inducible 14; Fgfrp2; fibroblast growth factor regulated protein 2; fibroblast growth factor-inducible immediate-early response protein 14; Fibroblast growth factor-regulated protein 2; FN14; HPIP; TNF receptor superfamily member 12A; TNFRSF12A; Tumor necrosis factor receptor superfamily member 12A; tumor necrosis factor receptor superfamily, member 12A; TweakR; TWEAK-R; Tweak-receptor; type I transmembrane protein Fn14 | |
Rat | |
Protein A/G | |
RUO | |
27279 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |