missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TWIST1 Polyclonal Antibody, FITC, Invitrogen™
Sheep Polyclonal Antibody
Marke: Thermo Scientific PA546889
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
In direct ELISAs, less than 16% cross-reactivity with recombinant human Twist-2 is observed.
TWIST1 Polyclonal antibody specifically detects TWIST1 in Human samples. It is validated for Flow Cytometry.Spezifikation
TWIST1 | |
Polyclonal | |
Buffered saline solution with BSA and 0.09% sodium azide | |
Q15672 | |
TWIST1 | |
E. coli-derived recombinant human Twist-1 Met1-His202 | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG |
Flow Cytometry | |
FITC | |
TWIST1 | |
AA960487; ACS3; B-HLH DNA binding protein; bHLHa38; BPES2; BPES3; charlie chaplin; class A basic helix-loop-helix protein 38; CRS; CRS1; CSO; H-twist; M-twist; p ACS3; Pde; pdt; Pluridigite; polydactyly EMS; SCS; Ska<m10Jus>; Ska<sup>m10Jus</sup>; Ska10; Twist; twist basic helix-loop-helix transcription factor 1; twist family bHLH transcription factor 1; twist gene homolog 1; twist homolog 1; TWIST homolog of drosophila; Twist1; twist-related protein 1 | |
Sheep | |
Antigen affinity chromatography | |
RUO | |
7291 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |