missing translation for 'onlineSavingsMsg'
Learn More

TWIST1 Polyclonal Antibody, Invitrogen™

TWIST1 Antibody, PA5-41422, from Invitrogen™. Species Reactivity: Bovine, Canine, Equine, Guinea Pig, Human, Mouse, Porcine, Rabbit, Rat, Zebrafish; Applications: IHC, WB.

Marke:  Invitrogen PA541422

Weitere Details : Netto-Gewicht : 0.00010kg

Artikelnummer. 15544792

Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen
Alternative Produkte anzeigen »


Beschreibung

Beschreibung

TWIST1 Antibody, PA5-41422, from Invitrogen. Species Reactivity: Bovine, Canine, Equine, Guinea Pig, Human, Mouse, Porcine, Rabbit, Rat, Zebrafish; Applications: Immunohistochemistry, Western Blot.

Basic helix-loop-helix (bHLH) transcription factors have been implicated in cell lineage determination and differentiation. The protein encoded by this gene is a bHLH transcription factor and shares similarity with another bHLH transcription factor, Dermo1. The strongest expression of this mRNA is in placental tissue; in adults, mesodermally derived tissues express this mRNA preferentially. Mutations in this gene have been found in patients with Saethre-Chotzen syndrome.
Host Species: Rabbit
Cross Reactivity: Bovine, Canine, Equine, Guinea Pig, Human, Mouse, Porcine, Rabbit, Rat, Zebrafish
Isotype: IgG
Immunogen: Synthetic peptide directed towards the C-terminal of human TWIST1
Formulation: 100μL purified antibody (1mg/mL) in PBS with 2% sucrose and 0.09% sodium azide
Synonyms: Twist homolog 1 (Drosophila), SCS, ACS3, CRS1, BPES2, BPES3, TWIST, bHLHa38
Uniprot Accession: P26687, Q15672
Entrez GeneID: 22160, 7291
Gene Symbol: TWIST1
Peptide sequence: DKLSKIQTLK LAARYIDFLY QVLQSDELDS KMASCSYVAH ERLSYAFSVW Sequence homology: Cow: 100%; Dog: 79%; Guinea Pig: 79%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 79%; Rat: 100%; Zebrafish: 100%
Spezifikation

Spezifikation

TWIST1
Polyclonal
100μL purified antibody (1mg/mL) in PBS with 2% sucrose and 0.09% sodium azide
Twist homolog 1 (Drosophila), SCS, ACS3, CRS1, BPES2, BPES3, TWIST, bHLHa38
Rabbit
100 μL
22160, 7291
Store at 2°-8°C for up to 1 month. For long term storage, store at -20°C
IgG
Immunohistochemistry, Western Blot
Unconjugated
P26687, Q15672
TWIST1
Synthetic peptide directed towards the C-terminal of human TWIST1
Primary
Bovine, Canine, Equine, Guinea Pig, Human, Mouse, Porcine, Rabbit, Rat, Zebrafish
Purified
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote