missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TXK Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA514797
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
TXK Polyclonal antibody specifically detects TXK in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
TXK | |
Polyclonal | |
Unconjugated | |
TXK | |
A130089B16Rik; BTKL; Protein-tyrosine kinase 4; PSCTK5; PTK4; PTK4 protein tyrosine kinase 4; PTK-RL-18; resting lymphocyte kinase; Rlk; TKL; TXK; TXK tyrosine kinase; tyrosine kinase; tyrosine-protein kinase TXK | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
7294 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
P42681 | |
TXK | |
KLH conjugated synthetic peptide between 22-52 amino acids from the N-terminal region of human TXK | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |