missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TXLNB Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA533095
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Dog, Bovine, Bat, Elephant, Panda, Rabbit (100%) Mouse, Rat, Hamster (94%).
TXLNB Polyclonal antibody specifically detects TXLNB in Canine, Human, Non-human Primate, Rabbit samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
TXLNB | |
Polyclonal | |
Unconjugated | |
TXLNB | |
2310001N14Rik; beta-taxilin; C6orf198; dJ522B19.2; hMDP77; LST001; Mdp77; muscle specific protein; muscle-derived protein 77; RGD1307979; RP3-522B19.2; taxilin beta; Txlnb | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100346890, 167838, 476224 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q8N3L3 | |
TXLNB | |
Synthetic 17 amino acid peptide from internal region of human TXLNB. | |
50 μg | |
Primary | |
Canine, Human, Non-human Primate, Rabbit | |
Antibody | |
IgG |