missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TXNDC12 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA524798
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
TXNDC12 Polyclonal antibody specifically detects TXNDC12 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
TXNDC12 | |
Polyclonal | |
Unconjugated | |
Txndc12 | |
0610040B21Rik; AG1; AGR1; anterior gradient homolog 1; endoplasmic reticulum protein ERp19; endoplasmic reticulum resident protein 18; Endoplasmic reticulum resident protein 19; endoplasmic reticulum thioredoxin superfamily member, 18 kDa; ER protein 18; ER protein 19; ERp16; ERP18; ERP19; hAG-1; hTLP19; ortholog of endoplasmic reticulum thioredoxin superfamily member, 18 kDa TLP19; PDIA16; protein disulfide isomerase family A, member 16; RGD1305960; thioredoxin domain containing 12; thioredoxin domain containing 12 (endoplasmic reticulum); thioredoxin domain-containing protein 12; Thioredoxin-like protein p19; TLP19; TXNDC12; UNQ713/PRO1376 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
51060, 66073 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.25 mg/mL | |
PBS with 0.09% sodium azide | |
O95881, Q9CQU0 | |
Txndc12 | |
KLH conjugated synthetic peptide between 144-172 amino acids from the C-terminal region of human TXNDC12 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |