missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMX4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA543385
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: GVDEERSEAN DQGPPGEDGV TREEVEPEEA EEGISEQPCP ADTEVVEDSL Sequence homology: Human: 100%; Pig: 79%
TMX4 Polyclonal antibody specifically detects TMX4 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
TMX4 | |
Polyclonal | |
Unconjugated | |
TMX4 | |
2810417D04Rik; 4930500L08Rik; AI843224; AW046784; D2Bwg1356e; DJ971N18.2; Kiaa1162; mKIAA1162; PDIA14; protein disulfide isomerase family A, member 14; PSEC0095; RGD1305146; thioredoxin domain containing 13; thioredoxin domain-containing protein 13; thioredoxin related transmembrane protein 4; thioredoxin-related transmembrane protein 4; Tmx4; Txndc13; UNQ475/PRO938 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
56255 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9H1E5 | |
TMX4 | |
Synthetic peptide directed towards the C-terminal of human TXNDC13 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |