missing translation for 'onlineSavingsMsg'
Learn More

ERP44 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA528484

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 13472597

  • 474.00 € / 100 Mikroliter
Verfügbar ab: 22-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (98%), Rat (99%), Zebrafish (82%), Xenopus laevis (83%), Pig (97%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

ERP44 Polyclonal antibody specifically detects ERP44 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.
Spezifikation

Spezifikation

ERP44
Polyclonal
Unconjugated
ERP44
1110001E24Rik; AI849526; AL033348; BWK4; endoplasmic reticulum protein 44; endoplasmic reticulum resident protein 44; endoplasmic reticulum resident protein 44 kDa; endoplasmic reticulum resident protein 44kDa; endoplasmic reticulum resident protein ERp44; ER protein 44; Erp44; KIAA0573; OTTHUMP00000063799; PDIA10; protein disulfide isomerase family A, member 10; thioredoxin domain containing 4 (endoplasmic reticulum); thioredoxin domain-containing protein 4; Txndc4; UNQ532/PRO1075
Rabbit
Antigen affinity chromatography
RUO
23071, 76299
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q9BS26, Q9D1Q6
ERP44
Recombinant fragment corresponding to a region within amino acids 204 and 406 of Human TXNDC4
100 μL
Primary
Human, Mouse
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote