Learn More
TXNDC5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA528520
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: 293T, HeLa, HepG2, Huh-7. Predicted reactivity: Mouse (83%), Rat (84%), Bovine (85%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TXNDC5 Polyclonal antibody specifically detects TXNDC5 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
TXNDC5 | |
Polyclonal | |
Unconjugated | |
Txndc5 | |
AL022641; ENDOPDI; endoplasmic reticulum protein ERp46; endoplasmic reticulum resident protein 46; endothelial protein disulphide isomerase; ER protein 46; ERp46; FLJ21353; FLJ90810; HCC2; HCC-2; MGC3178; PC-TRP; PDIA15; Plasma cell-specific thioredoxin-related protein; protein disulfide isomerase family A, member 15; STRF8; thiore; thioredoxin domain containing 5; thioredoxin domain containing 5 (endoplasmic reticulum); thioredoxin domain-containing protein 5; thioredoxin related protein; thioredoxin-like protein p46; Tlp46; TXNDC5; UNQ364; UNQ364/PRO700 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
81567 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8NBS9 | |
Txndc5 | |
Recombinant fragment corresponding to a region within amino acids 198 and 432 of Human TXNDC5 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |