missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TXNDC6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA530945
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: NT2D1, IMR32, U87-MG. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TXNDC6 Polyclonal antibody specifically detects TXNDC6 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
TXNDC6 | |
Polyclonal | |
Unconjugated | |
NME9 | |
MGC129586; NM23-H9; NME gene family member 9; NME/NM23 family member 9; NME9; Thioredoxin domain-containing protein 6; thioredoxin-like 2; thioredoxin-like protein 2; TXL2; Txl-2; TXNDC6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
347736 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q86XW9 | |
NME9 | |
Recombinant protein fragment corresponding to a region within amino acids 1 and 142 of Human TXNDC6 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |