missing translation for 'onlineSavingsMsg'
Learn More

TXNL2 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA531160

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 13425677

  • 466.00 € / 100 Mikroliter
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Recommended positive controls: 293T, A431, Jurkat, Raji, mouse brain. Predicted reactivity: Mouse (96%), Rat (96%), Xenopus laevis (80%), Pig (94%), Chicken (86%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

TXNL2 Polyclonal antibody specifically detects TXNL2 in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.
Spezifikation

Spezifikation

TXNL2
Polyclonal
Unconjugated
GLRX3
bA500G10.4; FLJ11864; Glrx3; GLRX4; glutaredoxin 3; glutaredoxin 4; glutaredoxin-3; GRX3; GRX4; HUSSY-22; Picot; PKC interacting cousin of thioredoxin; PKC-interacting cousin of thioredoxin; PKCq-interacting protein; PKC-theta-interacting protein; testicular tissue protein Li 75; thioredoxin-like 2; thioredoxin-like protein 2; TXNL2; TXNL3
Rabbit
Antigen affinity chromatography
RUO
10539, 30926
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.61 mg/mL
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
O76003, Q9CQM9
GLRX3
Recombinant fragment corresponding to a region within amino acids 83 and 335 of Human TXNL2
100 μL
Primary
Human, Mouse
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote