Learn More
TXNL2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA531294
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: 293T, A431, Raji. Predicted reactivity: Mouse (92%), Rat (92%), Xenopus laevis (80%), Pig (90%), Chicken (83%), Bovine (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TXNL2 Polyclonal antibody specifically detects TXNL2 in Human samples. It is validated for Western Blot.Spezifikation
TXNL2 | |
Polyclonal | |
Unconjugated | |
GLRX3 | |
bA500G10.4; FLJ11864; Glrx3; GLRX4; glutaredoxin 3; glutaredoxin 4; glutaredoxin-3; GRX3; GRX4; HUSSY-22; Picot; PKC interacting cousin of thioredoxin; PKC-interacting cousin of thioredoxin; PKCq-interacting protein; PKC-theta-interacting protein; testicular tissue protein Li 75; thioredoxin-like 2; thioredoxin-like protein 2; TXNL2; TXNL3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10539 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.38 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
O76003 | |
GLRX3 | |
Recombinant fragment corresponding to a region within amino acids 1 and 241 of Human TXNL2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |