Learn More
TYRO3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA528682
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: TYRO3 tansfected 293T cell. Predicted reactivity: Mouse (91%), Rat (92%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TYRO3 Polyclonal antibody specifically detects TYRO3 in Human samples. It is validated for Western Blot.Spezifikation
TYRO3 | |
Polyclonal | |
Unconjugated | |
TYRO3 | |
AI323366; Brt; Bruton agammaglobulinemia tyrosine kinase; BYK; Dtk; Etk-2; etk2/tyro3; FLJ16467; Rek; Rse; Sky; soluble tyro 3; soluble tyro3; styro 3; styro3; TIF; TK19-2; tyro 3; TYRO3; TYRO3 protein tyrosine kinase; TYRO3 protein tyrosine kinase 3; tyrosine-protein kinase byk; Tyrosine-protein kinase DTK; tyrosine-protein kinase receptor TYRO3; Tyrosine-protein kinase RSE; tyrosine-protein kinase SKY; Tyrosine-protein kinase TIF | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7301 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.17 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q06418 | |
TYRO3 | |
Recombinant fragment corresponding to a region within amino acids 497 and 890 of Human TYRO3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |