missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tyrosinase Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA514232
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Tyrosinase Polyclonal antibody specifically detects Tyrosinase in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
Tyrosinase | |
Polyclonal | |
Unconjugated | |
TYR | |
albino; albino locus protein; ATN; C; CMM8; LB24-AB; monophenol monooxygenase; Oca1; OCA1A; OCAIA; oculocutaneous albinism IA; SHEP3; SK29-AB; skc35; Tumor rejection antigen AB; TYR; Tyrosinase; tyrosinase (oculocutaneous albinism IA); tyrosinase precursor | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
7299 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
PBS with 0.09% sodium azide | |
P14679 | |
TYR | |
KLH conjugated synthetic peptide between 486-513 amino acids from the C-terminal region of human Tyrosinase | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |